missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-fluoro-2-nitrobenzoate, 98%

Catalog No. AAH6130714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6130714 25 g
AAH6130706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6130714 Supplier Thermo Scientific Chemicals Supplier No. H6130714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 393-85-1 | C8H6FNO4 | 199.14 g/mol

Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 393-85-1
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD03425619
InChI Key LCAUVFVOLNRVKG-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorobenzoic acid methyl ester, 5-fluoro-2-nitrobenzoic acid methyl ester, 2-nitro-5-fluorobenzoic acid, methyl ester, benzoic acid, 5-fluoro-2-nitro-, methyl ester, methyl 5fluoro-2-nitrobenzoate, methyl5-fluoro-2-nitrobenzoate, 2-nitro-5-fluorobenzoic acid,methyl ester, 3-fluoro-6-nitrobenzoic acid methyl ester, 5-fluoro-2-nitro-benzoic acid methyl ester, benzoic acid,5-fluoro-2-nitro-, methyl ester
PubChem CID 5152049
IUPAC Name methyl 5-fluoro-2-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(F)=C1)[N+]([O-])=O

Specifications

Melting Point 35°C to 37°C
Color White
Quantity 25 g
Solubility Information Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.
Formula Weight 199.14
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 5-fluoro-2-nitrobenzoate
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only