missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate 98.0+%, TCI America™

Catalog No. p-7132946
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M19755G 5 g
M197525G 25 g
2 options

Catalog No. M19755G

Supplier: TCI America M19755G

Only null left

Chemical Identifiers

CAS 62140-67-4
Molecular Formula C11H14O5S
Molecular Weight (g/mol) 258.288
MDL Number MFCD01317547
InChI Key UUARTHQJSZTOEM-UHFFFAOYSA-N
Synonym 5-(Ethylsulfonyl)-2-methoxybenzoic Acid Methyl Ester
PubChem CID 6454351
IUPAC Name methyl 5-ethylsulfonyl-2-methoxybenzoate
SMILES CCS(=O)(=O)C1=CC(=C(C=C1)OC)C(=O)OC

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
Formula Weight 258.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate