missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Chloro-2-methoxybenzoate 97.0+%, TCI America™

Catalog No. C145725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C145725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C145725G Supplier TCI America Supplier No. C145725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33924-48-0
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017511
InChI Key HPTHYBXMNNGQEF-UHFFFAOYSA-N
Synonym methyl 5-chloro-o-anisate, methyl-5-chloro-2-methoxybenzoate, 5-chloro-2-methoxybenzoic acid methyl ester, benzoic acid, 5-chloro-2-methoxy-, methyl ester, 5-chloro-o-anisic acid methyl ester, 5-chloro-2-methoxy-benzoic acid methyl ester, pubchem3709, methyl5-chloro-2-methoxybenzoate, acmc-1agpb, dsstox_cid_31509
PubChem CID 118574
IUPAC Name methyl 5-chloro-2-methoxybenzoate
SMILES COC1=C(C=C(C=C1)Cl)C(=O)OC

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 g
Formula Weight 200.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Chloro-2-methoxybenzoate