missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-chloro-2-methoxybenzoate, 98%

Catalog No. p-7080999
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1078714 25 g
AAA1078722 100 g
AAA1078736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1078714 Supplier Thermo Scientific Chemicals Supplier No. A1078714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33924-48-0 | C9H9ClO3 | 200.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33924-48-0
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017511
InChI Key HPTHYBXMNNGQEF-UHFFFAOYSA-N
Synonym methyl 5-chloro-o-anisate, methyl-5-chloro-2-methoxybenzoate, 5-chloro-2-methoxybenzoic acid methyl ester, benzoic acid, 5-chloro-2-methoxy-, methyl ester, 5-chloro-o-anisic acid methyl ester, 5-chloro-2-methoxy-benzoic acid methyl ester, pubchem3709, methyl5-chloro-2-methoxybenzoate, acmc-1agpb, dsstox_cid_31509
PubChem CID 118574
IUPAC Name methyl 5-chloro-2-methoxybenzoate
SMILES COC1=C(C=C(C=C1)Cl)C(=O)OC

Specifications

Density 1.259
Boiling Point 235°C to 240°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 25 g
Beilstein 2719088
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material Methyl 5-chloro-2-methoxybenzoate
EINECSNumber 251-743-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only