missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Bromo-2-methylbenzoate 98.0+%, TCI America™

Catalog No. M25425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25425G 5 g
M25421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25425G Supplier TCI America Supplier No. M25425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79669-50-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD09839555
InChI Key FDCYLMYCHALQJR-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-methyl-benzoate, methyl 5-bromo-o-toluate, methyl5-bromo-2-methylbenzoate, 5-bromo-2-methyl-benzoic acid methyl ester, benzoic acid, 5-bromo-2-methyl-, methyl ester, 5-bromo-2-methylbenzoic acid methyl ester, 5-bromo-2-methylbenzoicacid, methyl 4-bromo-toluate, pubchem10936, acmc-1bhme
PubChem CID 316201
IUPAC Name methyl 5-bromo-2-methylbenzoate
SMILES CC1=C(C=C(C=C1)Br)C(=O)OC

Specifications

Melting Point 48°C
Color White
Boiling Point 265°C
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromo-2-methylbenzoate