missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Bromo-2-iodobenzoate 95.0+%, TCI America™
$31.68 - $117.43
Chemical Identifiers
| CAS | 181765-86-6 |
|---|---|
| Molecular Formula | C8H6BrIO2 |
| Molecular Weight (g/mol) | 340.94 |
| MDL Number | MFCD00144771 |
| InChI Key | CJRHLSZJEFJDLA-UHFFFAOYSA-N |
| Synonym | 5-Bromo-2-iodobenzoic Acid Methyl Ester |
| PubChem CID | 11078356 |
| IUPAC Name | methyl 5-bromo-2-iodobenzoate |
| SMILES | COC(=O)C1=C(I)C=CC(Br)=C1 |
Chemical Identifiers
| 181765-86-6 | |
| 340.94 | |
| CJRHLSZJEFJDLA-UHFFFAOYSA-N | |
| 11078356 | |
| COC(=O)C1=C(I)C=CC(Br)=C1 |
| C8H6BrIO2 | |
| MFCD00144771 | |
| 5-Bromo-2-iodobenzoic Acid Methyl Ester | |
| methyl 5-bromo-2-iodobenzoate |
Specifications
| 181765-86-6 | |
| White-Yellow | |
| C8H6BrIO2 | |
| 1 g | |
| CJRHLSZJEFJDLA-UHFFFAOYSA-N | |
| methyl 5-bromo-2-iodobenzoate | |
| 11078356 | |
| ≥95.0% (GC) | |
| Methyl 5-Bromo-2-iodobenzoate |
| 45°C | |
| 134°C | |
| MFCD00144771 | |
| 5-Bromo-2-iodobenzoic Acid Methyl Ester | |
| COC(=O)C1=C(I)C=CC(Br)=C1 | |
| 340.94 | |
| 340.94 | |
| Crystalline Powder |
Safety and Handling
TSCA : No