missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-chloronicotinate, 98%

CAS: 78686-79-0 | C7H5BrClNO2 | 250.476 g/mol

$199.48 - $707.89

Chemical Identifiers

CAS 78686-79-0
Molecular Formula C7H5BrClNO2
Molecular Weight (g/mol) 250.476
MDL Number MFCD03844848
InChI Key MOMQDEDQGJAKII-UHFFFAOYSA-N
PubChem CID 2763343
IUPAC Name methyl 5-bromo-2-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1Cl)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6479114
View Documents
Thermo Scientific Chemicals
H6479114
25 g
Each for $199.48
Only null left
 
AAH6479122
View Documents
Thermo Scientific Chemicals
H6479122
100 g
Each for $707.89
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78686-79-0
250.476
MOMQDEDQGJAKII-UHFFFAOYSA-N
methyl 5-bromo-2-chloropyridine-3-carboxylate
C7H5BrClNO2
MFCD03844848
2763343
COC(=O)C1=CC(=CN=C1Cl)Br

Specifications

78686-79-0
>110°C (230°F)
MFCD03844848
4394169
methyl 5-bromo-2-chloronicotinate, methyl5-bromo-2-chloronicotinate, 5-bromo-2-chloro-nicotinic acid methyl ester, methyl 5-bromo-2-chloro-3-pyridinecarboxylate, 5-bromo-2-chloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-chloro-, methyl ester, methyl 5-bromo-2-chloro-pyridine-3-carboxylate, 5-bromo-2-chloro-3-methoxycarbonyl pyridine, pubchem12966, acmc-209pf0
COC(=O)C1=CC(=CN=C1Cl)Br
250.476
250.48
Solid
48°C to 53°C
C7H5BrClNO2
25 g
Air, light, and moisture sensitive
MOMQDEDQGJAKII-UHFFFAOYSA-N
methyl 5-bromo-2-chloropyridine-3-carboxylate
2763343
98%
Methyl 5-bromo-2-chloronicotinate
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only