accessibility menu, dialog, popup

UserName

CH3OOBrNCl

CAS RN 78686-79-0

IUPAC Name: methyl 5-bromo-2-chloropyridine-3-carboxylate
Synonyms: methyl 5-bromo-2-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 250.48
Molecular Formula: C7H5BrClNO2
InChi Key: MOMQDEDQGJAKII-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CN=C1Cl
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)