missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Amino-2-chlorobenzoate 98.0+%, TCI America™

Catalog No. M28125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28125G 5 g
M28121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28125G Supplier TCI America Supplier No. M28125G

Chemical Identifiers

CAS 42122-75-8
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD07800315
InChI Key LBNPBOFVHYOPIB-UHFFFAOYSA-N
Synonym 5-Amino-2-chlorobenzoic Acid Methyl Ester
PubChem CID 5200412
IUPAC Name methyl 5-amino-2-chlorobenzoate
SMILES COC(=O)C1=C(Cl)C=CC(N)=C1

Specifications

Boiling Point 160°C
Quantity 5 g
Formula Weight 185.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 5-Amino-2-chlorobenzoate
TSCA No
Recommended Storage Refrigerator