missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-vinylbenzoate, 96%

Catalog No. AAH6005706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6005706 5 g
AAH6005703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6005706 Supplier Thermo Scientific Chemicals Supplier No. H6005706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1076-96-6 | C10H10O2 | 162.19 g/mol

Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1076-96-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00017218
InChI Key NUMHUJZXKZKUBN-UHFFFAOYSA-N
Synonym methyl 4-vinylbenzoate, methyl4-vinylbenzoate, methyl p-vinylbenzoate, methyl-4-vinylbenzoate, benzoic acid, 4-ethenyl-, methyl ester, benzoic acid, p-vinyl-, methyl ester, 4-vinyl benzoic acid methyl ester, acmc-1brgo, 4-vinyl-benzoic acid methyl ester
PubChem CID 583124
IUPAC Name methyl 4-ethenylbenzoate
SMILES COC(=O)C1=CC=C(C=C)C=C1

Specifications

Melting Point 32°C to 37°C
Quantity 5 g
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material Methyl 4-vinylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only