missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Nitrobenzoate 98.0+%, TCI America™

Catalog No. N0159500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0159500G 500 g
N015925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0159500G Supplier TCI America Supplier No. N0159500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-50-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007350
InChI Key YOJAHJGBFDPSDI-UHFFFAOYSA-N
Synonym methyl p-nitrobenzoate, benzoic acid, 4-nitro-, methyl ester, p-nitrobenzoic acid methyl ester, methyl-p-nitrobenzoate, methyl-4-nitrobenzoate, benzoic acid, p-nitro-, methyl ester, p-carbomethoxynitrobenzene, unii-nmn8cth864, nmn8cth864, 4-nitrobenzoic acid methyl ester
PubChem CID 12083
IUPAC Name methyl 4-nitrobenzoate
SMILES COC(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 96°C
Color Yellow
Quantity 500 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Nitrobenzoate
EINECSNumber (3)-1503
RTECSNumber DH5775000
TSCA Yes