missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(methylamino)benzoate, 98%

Catalog No. p-7021217
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1346006 5 g
AAA1346014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1346006 Supplier Thermo Scientific Chemicals Supplier No. A1346006
Only null left

CAS: 18358-63-9 | C9H11NO2 | 165.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is slightly soluble in water. (1.2 g/L) (25°C insoluble in hexane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 18358-63-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017198
InChI Key LLAMGYUWYUMHCH-UHFFFAOYSA-N
Synonym methyl 4-methylamino benzoate, methyl 4 methylamino benzoate, benzoic acid, 4-methylamino-, methyl ester, acmc-209eks, methyl 4-methylaninobenzoate, 4-carbomethoxy-n-methylaniline, methyl 4-methylamino-benzoate, methyl-4-methylaminobenzoate, 4-methoxycarbonyl-n-methylaniline, 4-methylamino-benzoic acid methylester
PubChem CID 140375
IUPAC Name methyl 4-(methylamino)benzoate
SMILES CNC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 92°C to 97°C
Quantity 5 g
Beilstein 2803327
Solubility Information It is slightly soluble in water. (1.2g/L) (25°C insoluble in hexane.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Methyl 4-(methylamino)benzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only