accessibility menu, dialog, popup

UserName

H3CNHOOCH3

CAS RN 18358-63-9

IUPAC Name: methyl 4-(methylamino)benzoate
Synonyms: methyl 4-(methylamino)benzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: LLAMGYUWYUMHCH-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results