missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxyphenylacetate 97.0+%, TCI America™

Catalog No. M133025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M133025G 25 g
M1330500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M133025G Supplier TCI America Supplier No. M133025G

Chemical Identifiers

CAS 23786-14-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00008454
InChI Key ZQYLDVNTWDEAJI-UHFFFAOYSA-N
Synonym methyl 4-methoxyphenylacetate, methyl 2-4-methoxyphenyl acetate, methyl 4-methoxyphenyl acetate, 4-methoxyphenylacetic acid methyl ester, 4-methoxy-phenyl-acetic acid methyl ester, methyl 4-methoxybenzeneacetate, methyl p-methoxyphenyl acetate, benzeneacetic acid, 4-methoxy-, methyl ester, acetic acid, p-methoxyphenyl-, methyl ester, methyl 2-p-methoxyphenyl acetate
PubChem CID 90266
IUPAC Name methyl 2-(4-methoxyphenyl)acetate
SMILES COC(=O)CC1=CC=C(OC)C=C1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
UN Number 1993
Formula Weight 180.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Methoxyphenylacetate