accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 23786-14-3

IUPAC Name: methyl 2-(4-methoxyphenyl)acetate
Synonyms: methyl 2-(4-methoxyphenyl)acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: ZQYLDVNTWDEAJI-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(OC)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)