Learn More
Methyl 4-methoxyphenylacetate, 97+%
CAS: 23786-14-3 | C10H12O3 | 180.20 g/mol
Supplier: Thermo Scientific Chemicals A1794036
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl 4-methoxyphenylacetate | |
| 1.135 | |
| >100°C (212°F) | |
| 1.516 | |
| 500 g | |
| methyl 4-methoxyphenylacetate, methyl 2-4-methoxyphenyl acetate, methyl 4-methoxyphenyl acetate, 4-methoxyphenylacetic acid methyl ester, 4-methoxy-phenyl-acetic acid methyl ester, methyl 4-methoxybenzeneacetate, methyl p-methoxyphenyl acetate, benzeneacetic acid, 4-methoxy-, methyl ester, acetic acid, p-methoxyphenyl-, methyl ester, methyl 2-p-methoxyphenyl acetate | |
| COC(=O)CC1=CC=C(OC)C=C1 | |
| 180.20 | |
| 180.23 |
| 23786-14-3 | |
| 157°C to 159°C (19 mmHg) | |
| C10H12O3 | |
| MFCD00008454 | |
| 2209665 | |
| ZQYLDVNTWDEAJI-UHFFFAOYSA-N | |
| methyl 2-(4-methoxyphenyl)acetate | |
| 90266 | |
| ≥97% |
Chemical Identifiers
| 23786-14-3 | |
| 180.20 | |
| ZQYLDVNTWDEAJI-UHFFFAOYSA-N | |
| 90266 | |
| COC(=O)CC1=CC=C(OC)C=C1 |
| C10H12O3 | |
| MFCD00008454 | |
| methyl 4-methoxyphenylacetate, methyl 2-4-methoxyphenyl acetate, methyl 4-methoxyphenyl acetate, 4-methoxyphenylacetic acid methyl ester, 4-methoxy-phenyl-acetic acid methyl ester, methyl 4-methoxybenzeneacetate, methyl p-methoxyphenyl acetate, benzeneacetic acid, 4-methoxy-, methyl ester, acetic acid, p-methoxyphenyl-, methyl ester, methyl 2-p-methoxyphenyl acetate | |
| methyl 2-(4-methoxyphenyl)acetate |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 245-886-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only