missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Methoxyacetoacetate 97.0+%, TCI America™
Supplier: TCI America M10315G
Specifications
| Methyl 4-Methoxyacetoacetate | |
| Yellow | |
| C6H10O4 | |
| 5 g | |
| QGBPKJFJAVDUNC-UHFFFAOYSA-N | |
| methyl 4-methoxy-3-oxobutanoate | |
| 123500 | |
| ≥97.0% (GC) |
| 41051-15-4 | |
| 113°C | |
| MFCD00010183 | |
| methyl 4-methoxyacetoacetate, methyl-4-methoxyacetoacetate, methyl 4-methoxy-3-oxobutyrate, butanoic acid, 4-methoxy-3-oxo-, methyl ester, methyl methoxyacetoacetate, 4-methoxyacetoacetic acid methyl ester, 4-methoxy-3-oxo-butyric acid methyl ester, pubchem10473, acmc-1aoxb, methyl4-methoxyacetoacetate | |
| COCC(=O)CC(=O)OC | |
| 146.142 | |
| 146.14 | |
| Liquid |
Chemical Identifiers
| 41051-15-4 | |
| 146.142 | |
| QGBPKJFJAVDUNC-UHFFFAOYSA-N | |
| methyl 4-methoxy-3-oxobutanoate |
| C6H10O4 | |
| MFCD00010183 | |
| 123500 | |
| COCC(=O)CC(=O)OC |
Safety and Handling
TSCA : Yes