missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxy-2-methylbenzoate 98.0+%, TCI America™

Catalog No. M28891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28891G 1 g
M28895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28891G Supplier TCI America Supplier No. M28891G

Chemical Identifiers

CAS 35598-05-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD06203918
InChI Key OGYAVWKYDVBIMW-UHFFFAOYSA-N
Synonym 4-Methoxy-2-methylbenzoic Acid Methyl Ester, Methyl 4-Methoxy-o-toluate, 4-Methoxy-o-toluic Acid Methyl Ester, Methyl 2-Methyl-p-anisate, 2-Methyl-p-anisic Acid Methyl Ester
PubChem CID 13257269
IUPAC Name methyl 4-methoxy-2-methylbenzoate
SMILES COC(=O)C1=C(C)C=C(OC)C=C1

Specifications

Color Yellow
Boiling Point 98°C
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Methoxy-2-methylbenzoate