missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Iodosalicylate 98.0+%, TCI America™
Supplier: TCI America M190825G
Specifications
| Methyl 4-Iodosalicylate | |
| 70°C | |
| MFCD06797864 | |
| methyl 4-iodosalicylate, 2-hydroxy-4-iodobenzoic acid methyl ester, benzoic acid,2-hydroxy-4-iodo-, methyl ester, methyl-4-iodosalicylate, methyl 4-iodosalicyclate, acmc-1c81e, methyl2-hydroxy-4-iodobenzoate, methyl 4-iodo-2-hydroxybenzoate, methyl-2-hydroxy-4-iodo-benzoate | |
| COC(=O)C1=C(C=C(C=C1)I)O | |
| 278.045 | |
| 278.05 | |
| Crystalline Powder |
| 18179-39-0 | |
| C8H7IO3 | |
| 25 g | |
| WUFUURSWOJROKY-UHFFFAOYSA-N | |
| methyl 2-hydroxy-4-iodobenzoate | |
| 11380407 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 18179-39-0 | |
| 278.045 | |
| WUFUURSWOJROKY-UHFFFAOYSA-N | |
| 11380407 | |
| COC(=O)C1=C(C=C(C=C1)I)O |
| C8H7IO3 | |
| MFCD06797864 | |
| methyl 4-iodosalicylate, 2-hydroxy-4-iodobenzoic acid methyl ester, benzoic acid,2-hydroxy-4-iodo-, methyl ester, methyl-4-iodosalicylate, methyl 4-iodosalicyclate, acmc-1c81e, methyl2-hydroxy-4-iodobenzoate, methyl 4-iodo-2-hydroxybenzoate, methyl-2-hydroxy-4-iodo-benzoate | |
| methyl 2-hydroxy-4-iodobenzoate |
Safety and Handling
TSCA : No