missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Iodosalicylate 98.0+%, TCI America™

Catalog No. M19085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19085G 5 g
M190825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19085G Supplier TCI America Supplier No. M19085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18179-39-0
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD06797864
InChI Key WUFUURSWOJROKY-UHFFFAOYSA-N
Synonym methyl 4-iodosalicylate, 2-hydroxy-4-iodobenzoic acid methyl ester, benzoic acid,2-hydroxy-4-iodo-, methyl ester, methyl-4-iodosalicylate, methyl 4-iodosalicyclate, acmc-1c81e, methyl2-hydroxy-4-iodobenzoate, methyl 4-iodo-2-hydroxybenzoate, methyl-2-hydroxy-4-iodo-benzoate
PubChem CID 11380407
IUPAC Name methyl 2-hydroxy-4-iodobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)I)O

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 278.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Iodosalicylate