missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodosalicylate, 98%

Catalog No. p-7028236
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2543806 5 g
AAB2543814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2543806 Supplier Thermo Scientific Chemicals Supplier No. B2543806
Only null left

CAS: 18179-39-0 | C8H7IO3 | 278.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18179-39-0
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD06797864
InChI Key WUFUURSWOJROKY-UHFFFAOYSA-N
Synonym methyl 4-iodosalicylate, 2-hydroxy-4-iodobenzoic acid methyl ester, benzoic acid,2-hydroxy-4-iodo-, methyl ester, methyl-4-iodosalicylate, methyl 4-iodosalicyclate, acmc-1c81e, methyl2-hydroxy-4-iodobenzoate, methyl 4-iodo-2-hydroxybenzoate, methyl-2-hydroxy-4-iodo-benzoate
PubChem CID 11380407
IUPAC Name methyl 2-hydroxy-4-iodobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)I)O

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
Sensitivity Light Sensitive
Formula Weight 278.05
Percent Purity 98%
Chemical Name or Material Methyl 4-iodosalicylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only