missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobenzoate, 98%

Catalog No. p-164312
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414460250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414460250 Supplier Thermo Scientific Chemicals Supplier No. 414460250

CAS: 619-44-3 | C8H7IO2 | 262.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-44-3
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
MDL Number MFCD00016353
InChI Key DYUWQWMXZHDZOR-UHFFFAOYSA-N
Synonym methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
PubChem CID 69273
IUPAC Name methyl 4-iodobenzoate
SMILES COC(=O)C1=CC=C(C=C1)I

Specifications

Melting Point 113°C to 117°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CO2CH3C6H4I
Quantity 25 g
Formula Weight 262.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-iodobenzoate, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
EINECSNumber 210-597-1
TSCA TSCA

RUO – Research Use Only