missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobenzoate, 98%

Catalog No. AAA1934218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1934218 50 g
AAA1934209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1934218 Supplier Thermo Scientific Chemicals Supplier No. A1934218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 619-44-3 | C8H7IO2 | 262.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-44-3
Molecular Formula C8H7IO2
MDL Number MFCD00016353
InChI Key DYUWQWMXZHDZOR-UHFFFAOYSA-N
Synonym methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
PubChem CID 69273
IUPAC Name methyl 4-iodobenzoate

Specifications

Melting Point 114°C to 116°C
Quantity 50 g
Sensitivity Light sensitive
SMILES COC(=O)C1=CC=C(C=C1)I
Molecular Weight (g/mol) 262.046
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material Methyl 4-iodobenzoate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 210-597-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only