missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodo-3-nitrobenzoate, 99%

CAS: 89976-27-2 | C8H6INO4 | 307.043 g/mol

$174.60 - $679.64

Chemical Identifiers

CAS 89976-27-2
Molecular Formula C8H6INO4
Molecular Weight (g/mol) 307.043
MDL Number MFCD08064223
InChI Key SCMBIQRYVKITCY-UHFFFAOYSA-N
PubChem CID 297884
IUPAC Name methyl 4-iodo-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2554406
View Documents
Thermo Scientific Chemicals
B2554406
5 g
Each for $174.60
Only null left
 
AAB2554414
View Documents
Thermo Scientific Chemicals
B2554414
25 g
Each for $679.64
Only null left
 
Description

Description

Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in DCM 10mg/0.5mL. Insoluble in water.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
Specifications

Chemical Identifiers

89976-27-2
307.043
SCMBIQRYVKITCY-UHFFFAOYSA-N
methyl 4-iodo-3-nitrobenzoate
C8H6INO4
MFCD08064223
297884
COC(=O)C1=CC(=C(C=C1)I)[N+](=O)[O-]

Specifications

89976-27-2
C8H6INO4
5 g
4-iodo-3-nitrobenzoic acid methyl ester, benzoic acid,4-iodo-3-nitro-, methyl ester, benzoic acid, 4-iodo-3-nitro-, methyl ester, methyl4-iodo-3-nitrobenzoate, acmc-20af7x, methyl-4-iodo-3-nitrobenzoate, 4-iodo-3-nitro benzoic methyl ester, 3-nitro-4-iodobenzoic acid methyl ester, 4-iodo-3-nitro-benzoic acid methyl ester
SCMBIQRYVKITCY-UHFFFAOYSA-N
methyl 4-iodo-3-nitrobenzoate
297884
99%
101°C to 104°C
MFCD08064223
Light sensitive
Soluble in DCM 10mg/0.5mL. Insoluble in water.
COC(=O)C1=CC(=C(C=C1)I)[N+](=O)[O-]
307.043
307.05
Methyl 4-iodo-3-nitrobenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only