missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodo-3-nitrobenzoate, 99%

Catalog No. AAB2554406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2554406 5 g
AAB2554414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2554406 Supplier Thermo Scientific Chemicals Supplier No. B2554406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89976-27-2 | C8H6INO4 | 307.043 g/mol

Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in DCM 10mg/0.5mL. Insoluble in water.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.

Chemical Identifiers

CAS 89976-27-2
Molecular Formula C8H6INO4
Molecular Weight (g/mol) 307.043
MDL Number MFCD08064223
InChI Key SCMBIQRYVKITCY-UHFFFAOYSA-N
PubChem CID 297884
IUPAC Name methyl 4-iodo-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 101°C to 104°C
Quantity 5 g
Sensitivity Light sensitive
Synonym 4-iodo-3-nitrobenzoic acid methyl ester, benzoic acid,4-iodo-3-nitro-, methyl ester, benzoic acid, 4-iodo-3-nitro-, methyl ester, methyl4-iodo-3-nitrobenzoate, acmc-20af7x, methyl-4-iodo-3-nitrobenzoate, 4-iodo-3-nitro benzoic methyl ester, 3-nitro-4-iodobenzoic acid methyl ester, 4-iodo-3-nitro-benzoic acid methyl ester
Solubility Information Soluble in DCM 10mg/0.5mL. Insoluble in water.
Formula Weight 307.05
Percent Purity 99%
Chemical Name or Material Methyl 4-iodo-3-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only