missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodo-2-methoxybenzoate, 98+%

Catalog No. AAB2552906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2552906 5 g
AAB2552914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2552906 Supplier Thermo Scientific Chemicals Supplier No. B2552906

CAS: 148490-97-5 | C9H9IO3 | 292.072 g/mol

Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light, oxidizing agents.

Chemical Identifiers

CAS 148490-97-5
Molecular Formula C9H9IO3
Molecular Weight (g/mol) 292.072
MDL Number MFCD06797865
InChI Key MCNOTXROWOGSGU-UHFFFAOYSA-N
Synonym benzoic acid,4-iodo-2-methoxy-, methyl ester, 4-iodo-2-methoxybenzoic acid methyl ester, 4-iodo-2-methoxy-benzoic acid methyl ester, pubchem4974, acmc-20adk9, methyl4-iodo-2-methoxybenzoate, methyl-4-iodo-2-methoxybenzoate, 2-methoxy-4-iodo-benzoic acid methyl ester
PubChem CID 22619468
IUPAC Name methyl 4-iodo-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1)I)C(=O)OC

Specifications

Melting Point 44°C to 46°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 292.07
Percent Purity ≥98%
Chemical Name or Material Methyl 4-iodo-2-methoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only