missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Hydroxyphenylacetate 98.0+%, TCI America™

Catalog No. H085625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H085625G 25 g
H0856500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H085625G Supplier TCI America Supplier No. H085625G

Chemical Identifiers

CAS 14199-15-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002387
InChI Key XGDZEDRBLVIUMX-UHFFFAOYSA-N
Synonym methyl 4-hydroxyphenylacetate, methyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methyl ester, benzeneacetic acid, 4-hydroxy-, methyl ester, methyl 4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid methl ester, acetic acid, p-hydroxyphenyl-, methyl ester, p-hydroxyphenylacetic acid methyl ester, 4-hydroxybenzeneacetic acid methyl ester, 4-hydroxy-phenyl-acetic acid methyl ester
PubChem CID 518900
ChEBI CHEBI:68078
IUPAC Name methyl 2-(4-hydroxyphenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)O

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 310°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Hydroxyphenylacetate
EINECSNumber (3)-3629
TSCA Yes