missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Hydroxymethyl)benzoate 98.0+%, TCI America™

Catalog No. H06405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06405G 5 g
H064025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06405G Supplier TCI America Supplier No. H06405G

Chemical Identifiers

CAS 6908-41-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00009984
InChI Key VBWFYEFYHJRJER-UHFFFAOYSA-N
Synonym methyl 4-hydroxymethyl benzoate, benzoic acid, 4-hydroxymethyl-, methyl ester, 4-carbomethoxybenzyl alcohol, 4-hydroxymethyl benzoic acid methyl ester, methyl p-hydroxymethyl benzoate, acmc-209o5w, methyl p-hydroxymethylbenzoate, methyl-4-hydroxymethylbenzoate, p-methoxycarbonylbenzyl alcohol
PubChem CID 81325
IUPAC Name methyl 4-(hydroxymethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)CO

Specifications

Melting Point 52°C
Color White
Boiling Point 157°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Hydroxymethyl)benzoate