missing translation for 'onlineSavingsMsg'
Learn More

Methyl (4-Hydroxybenzoyl)acetate 93.0+%, TCI America™

Catalog No. H08285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08285G 5 g
H08281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08285G Supplier TCI America Supplier No. H08285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32066-29-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00191516
InChI Key VZOFEVHSVUBEPH-UHFFFAOYSA-N
Synonym (4-Hydroxybenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester
PubChem CID 508465
IUPAC Name methyl 3-(4-hydroxyphenyl)-3-oxopropanoate
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (4-Hydroxybenzoyl)acetate