accessibility menu, dialog, popup

UserName

CAS RN 32066-29-8

IUPAC Name: methyl 3-(4-hydroxyphenyl)-3-oxopropanoate
Synonyms: methyl 3-(4-hydroxyphenyl)-3-oxopropanoate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: VZOFEVHSVUBEPH-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results