missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. p-7130415
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H021625G 25 g
H0216500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H021625G Supplier TCI America Supplier No. H021625G
Only null left

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002352
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 127°C
Color White
Boiling Point 280°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2450000
TSCA Yes