missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxybenzoate, 99%

Catalog No. p-7021803
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1428930 250 g
AAA142890B 1000 g
AAA142890I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1428930 Supplier Thermo Scientific Chemicals Supplier No. A1428930
Only null left

CAS: 99-76-3 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl 4-hydroxybenzoate is used as an anti-fungal agent. It is also used as a preservative in foods, beverages and cosmetics. It acts as an inhibitor of growth of molds and to lesser extent bacteria and as a vehicle for ophthalmic solution.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002352
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 125°C to 128°C
pH 5.8
Boiling Point ∼280°C (decomposition)
Odor Odorless
Quantity 250 g
Beilstein 509801
Merck Index 14,6107
Solubility Information Slightly soluble in water.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Methyl 4-hydroxybenzoate
Show More Show Less
EINECSNumber 202-785-7
RTECSNumber DH2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only