missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Hydroxy-3,5-dimethoxybenzoate 98.0+%, TCI America™

Catalog No. p-7133445
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28065G 5 g
M280625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28065G Supplier TCI America Supplier No. M28065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 884-35-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00017199
InChI Key ZMXJAEGJWHJMGX-UHFFFAOYSA-N
Synonym methyl syringate, methyl 3,5-dimethoxy-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3,5-dimethoxy-, methyl ester, syringic acid methyl ester, methyl 4-hydroxy-3,5-dimethoxy-benzoate, acmc-1bjig, ksc496o5n, syringic acid monomethyl ester, rarechem al bf 0102, timtec-bb sbb016976
PubChem CID 70164
ChEBI CHEBI:45820
IUPAC Name methyl 4-hydroxy-3,5-dimethoxybenzoate
SMILES COC1=CC(=CC(=C1O)OC)C(=O)OC

Specifications

Melting Point 108°C
Color White to Yellow
Quantity 5 g
Formula Weight 212.2
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Hydroxy-3,5-dimethoxybenzoate
EINECSNumber (3)-4633
TSCA No