accessibility menu, dialog, popup

UserName

CH3OOOCH3OHOH3C

CAS RN 884-35-5

IUPAC Name: methyl 4-hydroxy-3,5-dimethoxybenzoate
Synonyms: syringate methyl syringate
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: ZMXJAEGJWHJMGX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=C(O)C(OC)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)