missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxy-2-hexynoate, 97%

Catalog No. AAB2469006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2469006 5 g
AAB2469003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2469006 Supplier Thermo Scientific Chemicals Supplier No. B2469006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112780-04-8 | C7H10O3 | 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112780-04-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD04039770
InChI Key GMESNUCOCRRYJV-UHFFFAOYSA-N
Synonym methyl 4-hydroxy-2-hexynoate, 4-hydroxy-2-hexynoic acid methyl ester, acmc-20akge, methyl-4-hydroxy-2-hexynoate, 4-hydroxy-hex-2-ynoic acid methyl ester, 2-hexynoic acid,4-hydroxy-, methyl ester
PubChem CID 13859766
IUPAC Name methyl 4-hydroxyhex-2-ynoate
SMILES CCC(C#CC(=O)OC)O

Specifications

Refractive Index 1.462
Linear Formula CH3CH2CH(OH)C≡CCO2CH3
Quantity 5 g
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material Methyl 4-hydroxy-2-hexynoate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only