CAS RN 112780-04-8
IUPAC Name:
methyl 4-hydroxyhex-2-ynoate
Synonyms:
methyl 4-hydroxyhex-2-ynoate
Molecular Weight (g/mol):
142.15
Molecular Formula:
C7H10O3
InChi Key:
GMESNUCOCRRYJV-UHFFFAOYNA-N
SMILES:
CCC(O)C#CC(=O)OC
1
–
1
of
1
results
1
Promotions Available
Methyl 4-hydroxy-2-hexynoate, 97%
Methyl 4-hydroxy-2-hexynoate, 97%
CAS: 112780-04-8 Molecular Formula: C7H10O3 Molecular Weight (g/mol): 142.154 MDL Number: MFCD04039770 InChI Key: GMESNUCOCRRYJV-UHFFFAOYSA-N Synonym: methyl 4-hydroxy-2-hexynoate,4-hydroxy-2-hexynoic acid methyl ester,acmc-20akge,methyl-4-hydroxy-2-hexynoate,4-hydroxy-hex-2-ynoic acid methyl ester,2-hexynoic acid,4-hydroxy-, methyl ester PubChem CID: 13859766 IUPAC Name: methyl 4-hydroxyhex-2-ynoate SMILES: CCC(C#CC(=O)OC)O