missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Fluoro-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. M25351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25351G 1 g
M25355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25351G Supplier TCI America Supplier No. M25351G

Chemical Identifiers

CAS 329-59-9
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD00196161
InChI Key CNJJSTPBUHAEFH-UHFFFAOYSA-N
Synonym methyl 4-fluoro-3-nitro-benzoate, 4-fluoro-3-nitrobenzoic acid methyl ester, 4-fluoro-3-nitro-benzoic acid methyl ester, benzoic acid, 4-fluoro-3-nitro-, methyl ester, pubchem2045, acmc-209hxf, methyl 4-fluoro-3 nitrobenzoate, 4-carbomethoxy-2-nitrofluorobenzene, methyl4-fluoro-3-nitrobenzoate
PubChem CID 5219721
IUPAC Name methyl 4-fluoro-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O

Specifications

Melting Point 61°C
Color Yellow
Boiling Point 119°C
Quantity 1 g
Formula Weight 199.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Fluoro-3-nitrobenzoate