missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Cyanobenzoate 98.0+%, TCI America™
$141.67 - $812.73
Chemical Identifiers
| CAS | 1129-35-7 |
|---|---|
| Molecular Formula | C9H7NO2 |
| Molecular Weight (g/mol) | 161.16 |
| MDL Number | MFCD00001823 |
| InChI Key | KKZMIDYKRKGJHG-UHFFFAOYSA-N |
| Synonym | 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i |
| PubChem CID | 70791 |
| IUPAC Name | methyl 4-cyanobenzoate |
| SMILES | COC(=O)C1=CC=C(C=C1)C#N |
Chemical Identifiers
| 1129-35-7 | |
| 161.16 | |
| KKZMIDYKRKGJHG-UHFFFAOYSA-N | |
| 70791 | |
| COC(=O)C1=CC=C(C=C1)C#N |
| C9H7NO2 | |
| MFCD00001823 | |
| 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i | |
| methyl 4-cyanobenzoate |
Specifications
| 1129-35-7 | |
| White-Yellow | |
| C9H7NO2 | |
| 25 g | |
| KKZMIDYKRKGJHG-UHFFFAOYSA-N | |
| methyl 4-cyanobenzoate | |
| 70791 | |
| ≥98.0% (GC) | |
| Methyl 4-Cyanobenzoate |
| 68°C | |
| 142°C | |
| MFCD00001823 | |
| 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i | |
| COC(=O)C1=CC=C(C=C1)C#N | |
| 161.16 | |
| 161.16 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0527
TSCA : No