missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Cyanobenzoate 98.0+%, TCI America™

Catalog No. C111925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C111925G 25 g
C1119250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C111925G Supplier TCI America Supplier No. C111925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1129-35-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00001823
InChI Key KKZMIDYKRKGJHG-UHFFFAOYSA-N
Synonym 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i
PubChem CID 70791
IUPAC Name methyl 4-cyanobenzoate
SMILES COC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 68°C
Color White-Yellow
Boiling Point 142°C
Quantity 25 g
Formula Weight 161.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Cyanobenzoate
EINECSNumber (9)-0527
TSCA No