missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-(Chloromethyl)benzoate 98.0+%, TCI America™
$139.81 - $395.17
Chemical Identifiers
| CAS | 34040-64-7 |
|---|---|
| Molecular Formula | C9H9ClO2 |
| Molecular Weight (g/mol) | 184.619 |
| MDL Number | MFCD00511224 |
| InChI Key | SATDLKYRVXFXRE-UHFFFAOYSA-N |
| Synonym | methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester |
| PubChem CID | 36664 |
| IUPAC Name | methyl 4-(chloromethyl)benzoate |
| SMILES | COC(=O)C1=CC=C(C=C1)CCl |
Chemical Identifiers
| 34040-64-7 | |
| 184.619 | |
| SATDLKYRVXFXRE-UHFFFAOYSA-N | |
| 36664 | |
| COC(=O)C1=CC=C(C=C1)CCl |
| C9H9ClO2 | |
| MFCD00511224 | |
| methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester | |
| methyl 4-(chloromethyl)benzoate |
Specifications
| 34040-64-7 | |
| White-Yellow | |
| C9H9ClO2 | |
| 5 g | |
| methyl 4-chloromethyl benzoate, 4-chloromethyl benzoic acid methyl ester, methyl-4-chloromethylbenzoate, benzoic acid, 4-chloromethyl-, methyl ester, 4-methoxycarbonyl benzyl chloride, methyl alpha-chloro-p-toluate, methyl p-chloromethyl benzoate, p-methoxycarbonyl benzyl chloride, methyl a-chloro-p-toluate, p-toluic acid, .alpha.-chloro-, methyl ester | |
| COC(=O)C1=CC=C(C=C1)CCl | |
| 184.619 | |
| 184.62 | |
| Crystalline Powder |
| 39°C | |
| 165°C | |
| MFCD00511224 | |
| 3261 | |
| SATDLKYRVXFXRE-UHFFFAOYSA-N | |
| methyl 4-(chloromethyl)benzoate | |
| 36664 | |
| ≥98.0% (GC) | |
| Methyl 4-(Chloromethyl)benzoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator