missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chlorobutyrate 98.0+%, TCI America™

Catalog No. C082325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C082325ML Supplier TCI America Supplier No. C082325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3153-37-5
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00001003
InChI Key ZZUYIRISBMWFMV-UHFFFAOYSA-N
Synonym methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate
PubChem CID 76612
IUPAC Name methyl 4-chlorobutanoate
SMILES COC(=O)CCCCl

Specifications

Color Colorless
Boiling Point 174°C
Quantity 25 mL
UN Number 3272
Formula Weight 136.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Chlorobutyrate