missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Chloroacetoacetate 96.0+%, TCI America™
$88.56 - $506.33
Chemical Identifiers
| CAS | 32807-28-6 |
|---|---|
| Molecular Formula | C5H7ClO3 |
| Molecular Weight (g/mol) | 150.56 |
| MDL Number | MFCD00000938 |
| InChI Key | HFLMYYLFSNEOOT-UHFFFAOYSA-N |
| Synonym | methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 |
| PubChem CID | 36240 |
| IUPAC Name | methyl 4-chloro-3-oxobutanoate |
| SMILES | COC(=O)CC(=O)CCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C091025ML
|
TCI America
C091025ML |
25 mL |
Each for $88.56
|
|
|||||
|
C0910250ML
|
TCI America
C0910250ML |
250 mL |
Each for $506.33
|
|
|||||
Chemical Identifiers
| 32807-28-6 | |
| 150.56 | |
| HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
| 36240 | |
| COC(=O)CC(=O)CCl |
| C5H7ClO3 | |
| MFCD00000938 | |
| methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
| methyl 4-chloro-3-oxobutanoate |
Specifications
| 32807-28-6 | |
| Yellow | |
| C5H7ClO3 | |
| 25 mL | |
| methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
| COC(=O)CC(=O)CCl | |
| 150.56 | |
| 150.56 | |
| Liquid |
| 14°C | |
| 97°C | |
| MFCD00000938 | |
| 2922 | |
| HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
| methyl 4-chloro-3-oxobutanoate | |
| 36240 | |
| ≥96.0% (GC) | |
| Methyl 4-Chloroacetoacetate |
Safety and Handling
RTECSNumber : AK5115000
TSCA : Yes