missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Chloroacetoacetate 96.0+%, TCI America™
Supplier: TCI America C0910250ML
Specifications
| Methyl 4-Chloroacetoacetate | |
| 14°C | |
| 97°C | |
| MFCD00000938 | |
| 2922 | |
| HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
| methyl 4-chloro-3-oxobutanoate | |
| 36240 | |
| ≥96.0% (GC) |
| 32807-28-6 | |
| Yellow | |
| C5H7ClO3 | |
| 250 mL | |
| methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
| COC(=O)CC(=O)CCl | |
| 150.56 | |
| 150.56 | |
| Liquid |
Chemical Identifiers
| 32807-28-6 | |
| 150.56 | |
| HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
| 36240 | |
| COC(=O)CC(=O)CCl |
| C5H7ClO3 | |
| MFCD00000938 | |
| methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
| methyl 4-chloro-3-oxobutanoate |
Safety and Handling
RTECSNumber : AK5115000
TSCA : Yes