missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chloro-4-oxobutyrate 95.0+%, TCI America™

Catalog No. M285025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M285025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M285025G Supplier TCI America Supplier No. M285025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1490-25-1
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.558
MDL Number MFCD00000750
InChI Key SRXOJMOGPYFZKC-UHFFFAOYSA-N
Synonym methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
PubChem CID 73888
IUPAC Name methyl 4-chloro-4-oxobutanoate
SMILES COC(=O)CCC(=O)Cl

Specifications

Color Yellow
Boiling Point 65°C
Quantity 25 g
UN Number 3265
Formula Weight 150.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Chloro-4-oxobutyrate
EINECSNumber (2)-0918
TSCA Yes