missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chloro-3-methylbenzoate 98.0+%, TCI America™

Catalog No. M19355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19355G 5 g
M193525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19355G Supplier TCI America Supplier No. M19355G
Only null left

Chemical Identifiers

CAS 91367-05-4
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00151838
InChI Key QOTGNXMPQNGYDM-UHFFFAOYSA-N
Synonym 4-chloro-3-methylbenzoic acid methyl ester, benzoic acid, 4-chloro-3-methyl-, methyl ester, pubchem3710, acmc-209r8w, 2-chloro-5-methoxycarbonyl toluene, methyl 4-chloranyl-3-methyl-benzoate, 4-chloro-m-toluic acid methyl ester, 4-chloro-3-methylbenzoic acid, methyl ester
PubChem CID 7015786
IUPAC Name methyl 4-chloro-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 184.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Chloro-3-methylbenzoate