missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Bromopyrrole-2-carboxylate, TCI America™

$73.33 - $218.48

Chemical Identifiers

CAS 934-05-4
Molecular Formula C6H6BrNO2
Molecular Weight (g/mol) 204.023
MDL Number MFCD00832855
InChI Key ZXFCRVGOHJHZNF-UHFFFAOYSA-N
Synonym methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
PubChem CID 2763594
IUPAC Name methyl 4-bromo-1H-pyrrole-2-carboxylate
SMILES COC(=O)C1=CC(=CN1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25835G View Documents TCI America
M25835G
5g Each for $218.48
 
M25831G View Documents TCI America
M25831G
1g Each for $73.33
 
Specifications

Chemical Identifiers

934-05-4
204.023
ZXFCRVGOHJHZNF-UHFFFAOYSA-N
2763594
COC(=O)C1=CC(=CN1)Br
C6H6BrNO2
MFCD00832855
methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
methyl 4-bromo-1H-pyrrole-2-carboxylate

Specifications

White-Yellow
5g
C6H6BrNO2
methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
COC(=O)C1=CC(=CN1)Br
204.023
204.02
≥98.0% (GC)
103°C
934-05-4
MFCD00832855
ZXFCRVGOHJHZNF-UHFFFAOYSA-N
methyl 4-bromo-1H-pyrrole-2-carboxylate
2763594
Crystal-Powder at 20°C
Methyl 4-Bromopyrrole-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No