missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromopyrrole-2-carboxylate 98.0+%, TCI America™

Catalog No. M25831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25831G 1 g
M25835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25831G Supplier TCI America Supplier No. M25831G

Chemical Identifiers

CAS 934-05-4
Molecular Formula C6H6BrNO2
Molecular Weight (g/mol) 204.023
MDL Number MFCD00832855
InChI Key ZXFCRVGOHJHZNF-UHFFFAOYSA-N
Synonym methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
PubChem CID 2763594
IUPAC Name methyl 4-bromo-1H-pyrrole-2-carboxylate
SMILES COC(=O)C1=CC(=CN1)Br

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Bromopyrrole-2-carboxylate