missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromopyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH3391806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3391806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3391806 Supplier Thermo Scientific Chemicals Supplier No. H3391806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 934-05-4
Molecular Formula C6H6BrNO2
Molecular Weight (g/mol) 204.023
MDL Number MFCD00832855
InChI Key ZXFCRVGOHJHZNF-UHFFFAOYSA-N
Synonym methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
PubChem CID 2763594
IUPAC Name methyl 4-bromo-1H-pyrrole-2-carboxylate
SMILES COC(=O)C1=CC(=CN1)Br

Specifications

Melting Point 100°C to 102°C
Quantity 5 g
Formula Weight 204
Percent Purity 97%
Chemical Name or Material Methyl 4-bromopyrrole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only