missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Bromomethyl)benzoate 98.0+%, TCI America™

Catalog No. B205310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B205310G 10 g
B205325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B205310G Supplier TCI America Supplier No. B205310G

Chemical Identifiers

CAS 2417-72-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00032453
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
PubChem CID 256687
IUPAC Name methyl 4-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 56°C
Color White
Quantity 10 g
UN Number 3261
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Bromomethyl)benzoate