missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromocrotonate 80.0+%, TCI America™

Catalog No. M16895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16895G 5 g
M168925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16895G Supplier TCI America Supplier No. M16895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1117-71-1
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00000246,MFCD09998308
InChI Key RWIKCBHOVNDESJ-NSCUHMNNSA-N
Synonym methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester
PubChem CID 5369175
IUPAC Name methyl (2E)-4-bromobut-2-enoate
SMILES COC(=O)\C=C\CBr

Specifications

Boiling Point 75°C
Quantity 5 g
UN Number 1760
Formula Weight 179.01
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Bromocrotonate
RTECSNumber GQ3120000
TSCA Yes
Recommended Storage Refrigerator